Assay Details
| Gene Symbol: SYNGR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046436.2 | NP_001039901.1 | 4 - 5 | 659 | 78 | |
| GenBank mRNA | BC110272.1 | - | 4 - 5 | 662 | 78 | |
More Information
Set Membership: |
|

SYNGR4
Genomic Map