Assay Details
| Gene Symbol: DUSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046452.2 | NP_001039917.1 | 2 - 3 | 751 | 57 | |
| GenBank mRNA | BC105384.1 | - | 2 - 3 | 752 | 57 | |
| DQ473440.1 | - | 2 - 3 | 226 | 57 | ||
More Information
Set Membership: |
|

DUSP1
Genomic Map