Assay Details
| Gene Symbol: CSNK2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046454.1 | NP_001039919.1 | 6 - 7 | 683 | 67 | |
| GenBank mRNA | BC110170.1 | - | 6 - 7 | 683 | 67 | |
| HE864424.1 | - | 5 - 6 | 568 | 67 | ||
| HE864487.1 | - | 5 - 6 | 568 | 67 | ||
More Information
Set Membership: |
|

CSNK2B
Genomic Map