Assay Details
| Gene Symbol: OPRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046480.3 | NP_001039945.1 | 1 - 2 | 73 | 102 | |
| NM_001318764.1 | NP_001305693.1 | 1 - 2 | 73 | 102 | ||
| GenBank mRNA | BC112561.1 | - | 1 - 2 | 74 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112561.1, NM_001046480.3, NM_001318764.1 transcripts.
Set Membership: |
|

OPRK1
Genomic Map