Assay Details
| Gene Symbol: OPRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046480.3 | NP_001039945.1 | 2 - 3 | 372 | 84 | |
| NM_001318764.1 | NP_001305693.1 | 2 - 3 | 372 | 84 | ||
| GenBank mRNA | BC112561.1 | - | 2 - 3 | 373 | 84 | |
More Information
Set Membership: |
|

OPRK1
Genomic Map