Assay Details
| Gene Symbol: DNAJB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046503.1 | NP_001039968.1 | 3 - 4 | 864 | 101 | |
| GenBank mRNA | BC112518.1 | - | 3 - 4 | 864 | 101 | |
More Information
Set Membership: |
|

DNAJB4
Genomic Map