Assay Details
| Gene Symbol: PDZK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046521.2 | NP_001039986.1 | 3 - 4 | 400 | 74 | |
| GenBank mRNA | BC112562.1 | - | 3 - 4 | 401 | 74 | |
More Information
Set Membership: |
|

PDZK1IP1
Genomic Map