Assay Details
| Gene Symbol: RNF41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046525.1 | NP_001039990.1 | 6 - 7 | 859 | 60 | |
| GenBank mRNA | BC112724.1 | - | 6 - 7 | 859 | 60 | |
| BT030518.1 | - | 5 - 6 | 753 | 60 | ||
More Information
Set Membership: |
|

RNF41
Genomic Map