Assay Details
| Gene Symbol: BNIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046539.5 | NP_001040004.2 | 4 - 5 | 383 | 79 | |
| GenBank mRNA | BC110132.1 | - | 4 - 5 | 387 | 79 | |
| EE229689.1 | - | 4 - 5 | 384 | 79 | ||
More Information
Set Membership: |
|

BNIP1
Genomic Map