Assay Details
| Gene Symbol: HMGN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046547.1 | NP_001040012.1 | 1 - 2 | 216 | 128 | |
| GenBank mRNA | BC110182.1 | - | 1 - 2 | 216 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC110182.1, NM_001046547.1 transcripts.
Set Membership: |
|

HMGN4
Genomic Map