Assay Details
| Gene Symbol: CXCL10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046551.2 | NP_001040016.1 | 3 - 4 | 336 | 136 | |
| GenBank mRNA | BC112547.1 | - | 3 - 4 | 337 | 136 | |
| EU276062.1 | - | 3 - 4 | 284 | 136 | ||
More Information
Set Membership: |
|

CXCL10
Genomic Map