Assay Details
| Gene Symbol: GATC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046562.1 | NP_001040027.1 | 1 - 2 | 183 | 132 | |
| GenBank mRNA | BC112661.1 | - | 1 - 2 | 183 | 132 | |
More Information
Set Membership: |
|

GATC
Genomic Map