Assay Details
| Gene Symbol: NARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046572.2 | NP_001040037.1 | 11 - 12 | 1241 | 69 | |
| GenBank mRNA | BC105358.1 | - | 11 - 12 | 1242 | 69 | |
More Information
Set Membership: |
|

NARS1
Genomic Map