Assay Details
| Gene Symbol: NARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046572.2 | NP_001040037.1 | 14 - 15 | 1622 | 74 | |
| GenBank mRNA | BC105358.1 | - | 14 - 15 | 1623 | 74 | |
More Information
Set Membership: |
|

NARS1
Genomic Map