Assay Details
| Gene Symbol: NARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046572.2 | NP_001040037.1 | 1 - 2 | 119 | 114 | |
| GenBank mRNA | BC105358.1 | - | 1 - 2 | 120 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC105358.1, NM_001046572.2 transcripts.
Set Membership: |
|

NARS1
Genomic Map