Assay Details
| Gene Symbol: PUDP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046595.2 | NP_001040060.1 | 1 - 2 | 88 | 85 | |
| GenBank mRNA | BC105470.1 | - | 1 - 2 | 90 | 85 | |
More Information
Set Membership: |
|

PUDP
Genomic Map