Assay Details
| Gene Symbol: MRPS24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046601.2 | NP_001040066.1 | 1 - 2 | 54 | 83 | |
| GenBank mRNA | BC111632.1 | - | 1 - 2 | 55 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111632.1, NM_001046601.2 transcripts.
Set Membership: |
|

MRPS24
Genomic Map