Assay Details
| Gene Symbol: MAVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046620.2 | NP_001040085.1 | 6 - 7 | 1201 | 59 | |
| GenBank mRNA | BC112824.1 | - | 6 - 7 | 1202 | 59 | |
| KU695667.1 | - | 5 - 6 | 1177 | 59 | ||
More Information
Set Membership: |
|

MAVS
Genomic Map