Assay Details
| Gene Symbol: SEC11C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075175.2 | NP_001068643.1 | 2 - 3 | 270 | 57 | |
| GenBank mRNA | BC112783.1 | - | 2 - 3 | 271 | 57 | |
More Information
Set Membership: |
|

SEC11C
Genomic Map