Assay Details
| Gene Symbol: MRPL55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303490.1 | NP_001290419.1 | 2 - 3 | 257 | 76 | |
| NM_001303491.1 | NP_001290420.1 | 2 - 3 | 236 | 76 | ||
| NM_001303492.1 | NP_001290421.1 | 2 - 3 | 233 | 76 | ||
| NM_001303493.1 | NP_001290422.1 | 2 - 3 | 198 | 76 | ||
| NM_001303495.1 | NP_001290424.1 | 2 - 3 | 195 | 76 | ||
| GenBank mRNA | BC112839.1 | - | 2 - 3 | 226 | 76 | |
| BF040327.1 | - | 2 - 3 | 158 | 76 | ||
| BF044634.1 | - | 2 - 3 | 149 | 76 | ||
| DY461030.1 | - | 2 - 3 | 199 | 76 | ||
| EH126445.1 | - | 2 - 3 | 126 | 76 | ||
| EH156016.1 | - | 2 - 3 | 237 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112839.1 transcripts.
Set Membership: |
|

MRPL55
Genomic Map