Assay Details
| Gene Symbol: SEC61B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075292.2 | NP_001068760.1 | 1 - 2 | 310 | 92 | |
| GenBank mRNA | BC111652.1 | - | 1 - 2 | 311 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111652.1, NM_001075292.2 transcripts.
Set Membership: |
|

SEC61B
Genomic Map