Assay Details
| Gene Symbol: BCL2L11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075310.1 | NP_001068778.1 | 4 - 5 | 545 | 75 | |
| GenBank mRNA | BC110253.1 | - | 4 - 5 | 545 | 75 | |
| HM630308.1 | - | 2 - 3 | 461 | 75 | ||
More Information
Set Membership: |
|

BCL2L11
Genomic Map