Assay Details
| Gene Symbol: GOLT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075361.2 | NP_001068829.1 | 4 - 5 | 449 | 88 | |
| GenBank mRNA | BC111610.1 | - | 4 - 5 | 450 | 88 | |
More Information
Set Membership: |
|

GOLT1A
Genomic Map