Assay Details
| Gene Symbol: TMCO5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075609.2 | NP_001069077.1 | 8 - 9 | 601 | 86 | |
| GenBank mRNA | BC109751.1 | - | 8 - 9 | 602 | 86 | |
More Information
Set Membership: |
|

TMCO5B
Genomic Map