Assay Details
Gene Symbol: TEKT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075783.2 | NP_001069251.1 | 6 - 7 | 939 | 62 | |
GenBank mRNA | BC109833.1 | - | 6 - 7 | 940 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
