Assay Details
| Gene Symbol: NSMCE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075797.3 | NP_001069265.2 | 5 - 6 | 644 | 130 | |
| GenBank mRNA | BC109850.1 | - | 5 - 6 | 643 | 130 | |
| EV618408.1 | - | 5 - 6 | 645 | 130 | ||
More Information
Set Membership: |
|

NSMCE2
Genomic Map