Assay Details
| Gene Symbol: NXT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075805.1 | NP_001069273.1 | 1 - 2 | 264 | 94 | |
| GenBank mRNA | BC112489.1 | - | 1 - 2 | 264 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112489.1, NM_001075805.1 transcripts.
Set Membership: |
|

NXT1
Genomic Map