Assay Details
| Gene Symbol: SIL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075808.2 | NP_001069276.1 | 7 - 8 | 890 | 66 | |
| GenBank mRNA | BC109907.1 | - | 7 - 8 | 891 | 66 | |
More Information
Set Membership: |
|

SIL1
Genomic Map