Assay Details
| Gene Symbol: TSPAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075817.1 | NP_001069285.1 | 3 - 4 | 467 | 124 | |
| GenBank mRNA | BC109676.1 | - | 3 - 4 | 467 | 124 | |
More Information
Set Membership: |
|

TSPAN16
Genomic Map