Assay Details
| Gene Symbol: CPN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076080.2 | NP_001069548.1 | 7 - 8 | 1266 | 65 | |
| GenBank mRNA | BC105475.1 | - | 7 - 8 | 1267 | 65 | |
More Information
Set Membership: |
|

CPN1
Genomic Map