Assay Details
| Gene Symbol: ZBTB8OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076288.2 | NP_001069756.1 | 1 - 2 | 529 | 66 | |
| GenBank mRNA | BC110257.1 | - | 1 - 2 | 530 | 66 | |
More Information
Set Membership: |
|

ZBTB8OS
Genomic Map