Assay Details
| Gene Symbol: YEATS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076365.2 | NP_001069833.1 | 1 - 2 | 317 | 115 | |
| GenBank mRNA | BC109626.1 | - | 1 - 2 | 319 | 115 | |
More Information
Set Membership: |
|

YEATS4
Genomic Map