Assay Details
| Gene Symbol: DUSP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076374.1 | NP_001069842.1 | 2 - 3 | 305 | 78 | |
| GenBank mRNA | BC111276.1 | - | 2 - 3 | 305 | 78 | |
More Information
Set Membership: |
|

DUSP3
Genomic Map