Assay Details
| Gene Symbol: AQP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076378.2 | NP_001069846.1 | 4 - 5 | 536 | 72 | |
| GenBank mRNA | BC109728.1 | - | 4 - 5 | 537 | 72 | |
More Information
Set Membership: |
|

AQP7
Genomic Map