Assay Details
| Gene Symbol: DNAJC5G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076429.2 | NP_001069897.1 | 7 - 8 | 946 | 115 | |
| GenBank mRNA | BC111333.1 | - | 7 - 8 | 947 | 115 | |
More Information
Set Membership: |
|

DNAJC5G
Genomic Map