Assay Details
| Gene Symbol: LYPD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076442.1 | NP_001069910.1 | 2 - 3 | 203 | 57 | |
| GenBank mRNA | BC109635.1 | - | 2 - 3 | 203 | 57 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109635.1, NM_001076442.1 transcripts.
Set Membership: |
|

LYPD4
Genomic Map