Assay Details
| Gene Symbol: SCD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076945.1 | NP_001070413.1 | 1 - 2 | 503 | 55 | |
| GenBank mRNA | BC112711.1 | - | 1 - 2 | 503 | 55 | |
| EF014951.1 | - | 1 - 2 | 524 | 55 | ||
More Information
Set Membership: |
|

SCD5
Genomic Map