Assay Details
| Gene Symbol: KASH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076946.2 | NP_001070414.1 | 18 - 19 | 1667 | 107 | |
| GenBank mRNA | BC111305.1 | - | 18 - 19 | 1668 | 107 | |
More Information
Set Membership: |
|

KASH5
Genomic Map