Assay Details
| Gene Symbol: KASH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076946.2 | NP_001070414.1 | 1 - 2 | 138 | 78 | |
| GenBank mRNA | BC111305.1 | - | 1 - 2 | 139 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC111305.1, NM_001076946.2 transcripts.
Set Membership: |
|

KASH5
Genomic Map