Assay Details
| Gene Symbol: MAGEB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077050.2 | 1 | 85 | 64 | ||
| GenBank mRNA | BC109530.1 | - | 1 | 86 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109530.1, NM_001077050.2 transcripts.
Set Membership: |
|

MAGEB4
Genomic Map