Assay Details
| Gene Symbol: IQCF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077060.2 | NP_001070528.1 | 1 - 2 | 60 | 90 | |
| GenBank mRNA | BC109926.1 | - | 1 - 2 | 61 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109926.1, NM_001077060.2 transcripts.
Set Membership: |
|

IQCF5
Genomic Map