Assay Details
| Gene Symbol: FAM228A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077061.2 | NP_001070529.1 | 6 - 7 | 939 | 62 | |
| GenBank mRNA | BC109980.1 | - | 6 - 7 | 941 | 62 | |
More Information
Set Membership: |
|

FAM228A
Genomic Map