Assay Details
| Gene Symbol: SPINLW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077062.2 | NP_001070530.1 | 2 - 3 | 533 | 99 | |
| GenBank mRNA | BC109986.1 | - | 2 - 3 | 534 | 99 | |
More Information
Set Membership: |
|

SPINLW1
Genomic Map