Assay Details
| Gene Symbol: COG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077067.2 | NP_001070535.1 | 2 - 3 | 626 | 70 | |
| GenBank mRNA | BC110177.1 | - | 2 - 3 | 627 | 70 | |
More Information
Set Membership: |
|

COG8
Genomic Map