Assay Details
| Gene Symbol: C17H4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077088.2 | NP_001070556.1 | 3 - 4 | 477 | 76 | |
| GenBank mRNA | BC112545.1 | - | 3 - 4 | 478 | 76 | |
More Information
Set Membership: |
|

C17H4orf45
Genomic Map