Assay Details
| Gene Symbol: C17H4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077088.2 | NP_001070556.1 | 4 - 5 | 666 | 92 | |
| GenBank mRNA | BC112545.1 | - | 4 - 5 | 667 | 92 | |
More Information
Set Membership: |
|

C17H4orf45
Genomic Map