Assay Details
| Gene Symbol: DUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077873.2 | NP_001071341.1 | 85 | 113 | ||
| GenBank mRNA | BC111641.1 | - | 86 | 113 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DUT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077873.2 | NP_001071341.1 | 85 | 113 | ||
| GenBank mRNA | BC111641.1 | - | 86 | 113 | ||
Set Membership: |
|