Assay Details
| Gene Symbol: NUP42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077923.2 | NP_001071391.1 | 3 - 4 | 503 | 89 | |
| GenBank mRNA | BC110231.1 | - | 3 - 4 | 504 | 89 | |
More Information
Set Membership: |
|

NUP42
Genomic Map