Assay Details
| Gene Symbol: EGFL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078038.1 | NP_001071506.1 | 4 - 5 | 560 | 151 | |
| GenBank mRNA | BC111261.1 | - | 4 - 5 | 560 | 151 | |
More Information
Set Membership: |
|

EGFL7
Genomic Map