Assay Details
| Gene Symbol: CYSTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078164.1 | NP_001071632.1 | 2 - 3 | 331 | 105 | |
| GenBank mRNA | BC109536.1 | - | 2 - 3 | 331 | 105 | |
More Information
Set Membership: |
|

CYSTM1
Genomic Map